Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLEC11A
Entrez Gene ID
102131090
Gene Name
C-type lectin domain containing 11A
Gene Aliases
-
Chromosome Location
Chr.19: 51567913 - 51573136 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590028.2 XP_005590085.1 1 - 2 3183 135

View Full Product Details