Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM9C
Entrez Gene ID
102141431
Gene Name
family with sequence similarity 9 member C
Gene Aliases
-
Chromosome Location
Chr.X: 10773224 - 10782196 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005592975.2 XP_005593032.1 4 - 5 951 92

View Full Product Details