Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXA5
Entrez Gene ID
102137838
Gene Name
homeobox A5
Gene Aliases
-
Chromosome Location
Chr.3: 93582713 - 93586794 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005549935.2 XP_005549992.1 1 - 2 2561 90

View Full Product Details