Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PURB
Entrez Gene ID
102115217
Gene Name
purine rich element binding protein B
Gene Aliases
-
Chromosome Location
Chr.3: 75618666 - 75627663 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015447462.1 XP_015302948.1 1 - 1 808 69

View Full Product Details