Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC35B2
Entrez Gene ID
102131872
Gene Name
solute carrier family 35 member B2
Gene Aliases
-
Chromosome Location
Chr.4: 126265327 - 126268768 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005552861.2 XP_005552918.1 2 - 3 371 94

View Full Product Details