Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STEAP3
Entrez Gene ID
102135939
Gene Name
STEAP3 metalloreductase
Gene Aliases
-
Chromosome Location
Chr.12: 12924211 - 12963870 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005572950.2 XP_005573007.1 4 - 5 1293 80
XM_005572951.2 XP_005573008.1 4 - 5 2378 80

View Full Product Details