Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT8
Entrez Gene ID
102126411
Gene Name
nudix hydrolase 8
Gene Aliases
-
Chromosome Location
Chr.14: 6803534 - 6805547 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577051.2 XP_005577108.1 3 - 4 452 55

View Full Product Details