Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BSND
Entrez Gene ID
102143974
Gene Name
barttin CLCNK type accessory subunit beta
Gene Aliases
-
Chromosome Location
Chr.1: 172415176 - 172425901 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005543261.2 XP_005543318.1 1 - 2 456 75

View Full Product Details