Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUSD5
Entrez Gene ID
102117671
Gene Name
sushi domain containing 5
Gene Aliases
-
Chromosome Location
Chr.2: 14149646 - 14218244 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005545528.2 XP_005545585.1 3 - 4 561 86

View Full Product Details