Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MNX1
Entrez Gene ID
102131862
Gene Name
motor neuron and pancreas homeobox 1
Gene Aliases
-
Chromosome Location
Chr.3: 189928337 - 189934109 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015447368.1 XP_015302854.1 3 - 4 742 60

View Full Product Details