Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPS2
Entrez Gene ID
102133138
Gene Name
calcyphosine 2
Gene Aliases
-
Chromosome Location
Chr.11: 74484213 - 74550562 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015431216.1 XP_015286702.1 3 - 4 513 72
XM_015431217.1 XP_015286703.1 3 - 4 509 72
XM_015431218.1 XP_015286704.1 3 - 4 391 72
XM_015431219.1 XP_015286705.1 3 - 4 216 72
XM_015431220.1 XP_015286706.1 3 - 4 514 72

View Full Product Details