Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102125503
Entrez Gene ID
102125503
Gene Name
putative male-specific lethal-3 protein-like 2
Gene Aliases
MSL3P1
Chromosome Location
Chr.12: 123693743 - 123699643 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005574670.2 XP_005574727.1 2 - 3 1374 122

View Full Product Details