Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC39A1
Entrez Gene ID
102136645
Gene Name
solute carrier family 39 member 1
Gene Aliases
-
Chromosome Location
Chr.1: 97901387 - 97907103 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541732.2 XP_005541789.1 1 - 2 394 72

View Full Product Details