Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDLRAD1
Entrez Gene ID
102124514
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 173402335 - 173411815 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005543299.2 XP_005543356.1 6 - 7 467 103
XM_005543300.2 XP_005543357.1 5 - 6 338 103
XM_005543301.2 XP_005543358.1 3 - 4 200 103

View Full Product Details