Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNMT
Entrez Gene ID
102144850
Gene Name
phenylethanolamine N-methyltransferase
Gene Aliases
-
Chromosome Location
Chr.16: 51275257 - 51277870 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005584029.2 XP_005584086.1 2 - 3 726 61
XM_005584030.2 XP_005584087.1 2 - 3 728 61

View Full Product Details