Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1QTNF9
Entrez Gene ID
102116952
Gene Name
C1q and TNF related 9
Gene Aliases
-
Chromosome Location
Chr.17: 4622407 - 4645072 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005585487.2 XP_005585544.1 3 - 4 366 65
XM_015439071.1 XP_015294557.1 3 - 4 348 65

View Full Product Details