Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIRC1
Entrez Gene ID
102124593
Gene Name
disrupted in renal carcinoma 1
Gene Aliases
-
Chromosome Location
Chr.12: 77863528 - 77863834 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005596023.2 XP_005596080.1 1 - 1 78 63

View Full Product Details