Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HRNR
Entrez Gene ID
102140564
Gene Name
hornerin
Gene Aliases
-
Chromosome Location
Chr.1: 99683987 - 99698119 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541833.2 XP_005541890.1 9 - 10 6776 94

View Full Product Details