Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FABP3
Entrez Gene ID
102142791
Gene Name
fatty acid binding protein 3
Gene Aliases
-
Chromosome Location
Chr.1: 196661011 - 196668522 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005544157.2 XP_005544214.1 1 - 2 381 66

View Full Product Details