Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZSWIM3
Entrez Gene ID
102141138
Gene Name
zinc finger SWIM-type containing 3
Gene Aliases
-
Chromosome Location
Chr.10: 77527347 - 77549269 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319643.1 NP_001306572.1 1 - 2 349 81
GenBank mRNA AB172018.1 - 1 - 2 349 81

View Full Product Details