Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102138730
Entrez Gene ID
102138730
Gene Name
basic salivary proline-rich protein 3
Gene Aliases
PRB3
Chromosome Location
Chr.11: 11863015 - 11870368 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570157.2 XP_005570214.1 1 - 2 4174 88
XM_015430387.1 XP_015285873.1 1 - 2 4174 88

View Full Product Details