Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DEPDC4
Entrez Gene ID
102142847
Gene Name
DEP domain containing 4
Gene Aliases
-
Chromosome Location
Chr.11: 104018789 - 104040910 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015431421.1 XP_015286907.1 5 - 6 1190 131

View Full Product Details