Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLFN12L
Entrez Gene ID
102115313
Gene Name
schlafen family member 12 like
Gene Aliases
-
Chromosome Location
Chr.16: 31837291 - 31899584 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015438179.1 XP_015293665.1 4 - 5 1980 88

View Full Product Details