Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRTN
Entrez Gene ID
102144131
Gene Name
neurturin
Gene Aliases
-
Chromosome Location
Chr.19: 5949060 - 5971570 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005587660.2 XP_005587717.1 1 - 2 905 96
XM_015440891.1 XP_015296377.1 2 - 3 753 96
XM_015440892.1 XP_015296378.1 2 - 3 698 96

View Full Product Details