Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LILRA3
Entrez Gene ID
102128143
Gene Name
leukocyte immunoglobulin like receptor A3
Gene Aliases
-
Chromosome Location
Chr.19: 55195858 - 55201240 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590292.2 XP_005590349.2 2 - 3 321 79

View Full Product Details