Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BARHL2
Entrez Gene ID
102137917
Gene Name
BarH like homeobox 2
Gene Aliases
-
Chromosome Location
Chr.1: 136557494 - 136563534 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005542780.2 XP_005542837.1 1 - 2 982 62

View Full Product Details