Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102132156
Entrez Gene ID
102132156
Gene Name
complement C4-B
Gene Aliases
C4B
Chromosome Location
Chr.4: 138859330 - 138873551 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005553435.2 XP_005553492.1 21 - 22 2882 72

View Full Product Details