Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRXCR2
Entrez Gene ID
102119501
Gene Name
glutaredoxin and cysteine rich domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 145115045 - 145129169 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558135.2 XP_005558192.1 1 - 2 463 100

View Full Product Details