Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TEX22
Entrez Gene ID
102128994
Gene Name
testis expressed 22
Gene Aliases
-
Chromosome Location
Chr.7: 170772536 - 170788804 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005562410.2 XP_005562467.1 3 - 4 843 94
XM_015454278.1 XP_015309764.1 2 - 3 752 94

View Full Product Details