Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNFRSF10D
Entrez Gene ID
102136544
Gene Name
TNF receptor superfamily member 10d
Gene Aliases
-
Chromosome Location
Chr.8: 23385418 - 23414894 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005562837.2 XP_005562894.1 3 - 4 511 66

View Full Product Details