Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRFN4
Entrez Gene ID
102146195
Gene Name
leucine rich repeat and fibronectin type III domain containing 4
Gene Aliases
-
Chromosome Location
Chr.14: 7613698 - 7616728 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577109.2 XP_005577166.1 1 - 2 1652 77

View Full Product Details