Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMOD1
Entrez Gene ID
102144951
Gene Name
leiomodin 1
Gene Aliases
-
Chromosome Location
Chr.1: 60273999 - 60338036 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005540413.2 XP_005540470.1 4 - 5 1739 76

View Full Product Details