Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL19
Entrez Gene ID
102133068
Gene Name
interleukin 19
Gene Aliases
-
Chromosome Location
Chr.1: 65406047 - 65420236 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015449179.1 XP_015304665.1 6 - 7 1366 85

View Full Product Details