Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMOD2
Entrez Gene ID
102118699
Gene Name
leiomodin 2
Gene Aliases
-
Chromosome Location
Chr.3: 156465148 - 156473556 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005550645.1 XP_005550702.1 1 - 2 423 102

View Full Product Details