Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRN4
Entrez Gene ID
102115211
Gene Name
leucine rich repeat neuronal 4
Gene Aliases
-
Chromosome Location
Chr.10: 56441703 - 56453183 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005568389.2 XP_005568446.1 4 - 5 1330 61

View Full Product Details