Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCNT7
Entrez Gene ID
102125955
Gene Name
glucosaminyl (N-acetyl) transferase family member 7
Gene Aliases
-
Chromosome Location
Chr.10: 88338908 - 88345653 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015429989.1 XP_015285475.1 2 - 3 1060 78

View Full Product Details