Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTP4
Entrez Gene ID
102117670
Gene Name
receptor transporter protein 4
Gene Aliases
-
Chromosome Location
Chr.2: 10095995 - 10099460 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005545443.2 XP_005545500.1 1 - 2 502 60

View Full Product Details