Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PP2D1
Entrez Gene ID
102144725
Gene Name
protein phosphatase 2C like domain containing 1
Gene Aliases
-
Chromosome Location
Chr.2: 27582589 - 27607942 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005545671.2 XP_005545728.1 2 - 3 1329 81
GenBank mRNA AB168689.1 - 2 - 3 1272 81

View Full Product Details