Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL3
Entrez Gene ID
102118362
Gene Name
interleukin 3
Gene Aliases
-
Chromosome Location
Chr.6: 131053544 - 131055616 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005557713.2 XP_005557770.2 3 - 4 325 105

View Full Product Details