Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFR1
Entrez Gene ID
102115199
Gene Name
SWI5 dependent homologous recombination repair protein 1
Gene Aliases
-
Chromosome Location
Chr.9: 103197729 - 103202921 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566364.2 XP_005566421.1 2 - 3 609 121
XM_005566365.2 XP_005566422.1 2 - 3 240 121
XM_005566366.2 XP_005566423.1 2 - 3 315 121
XM_015456649.1 XP_015312135.1 2 - 3 208 121
XM_015456650.1 XP_015312136.1 2 - 3 205 121

View Full Product Details