Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCEMP1
Entrez Gene ID
102116476
Gene Name
mast cell expressed membrane protein 1
Gene Aliases
C19H19orf59
Chromosome Location
Chr.19: 8021074 - 8024309 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005587773.2 XP_005587830.1 5 - 6 581 88
XM_005587774.2 XP_005587831.1 5 - 6 923 88

View Full Product Details