Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC9
Entrez Gene ID
102118310
Gene Name
sialic acid binding Ig like lectin 9
Gene Aliases
SIGLEC7
Chromosome Location
Chr.19: 51962263 - 51974958 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590083.2 XP_005590140.1 6 - 7 1721 72

View Full Product Details