Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102130068
Entrez Gene ID
102130068
Gene Name
mesothelin-like protein
Gene Aliases
MSLNL
Chromosome Location
Chr.20: 797595 - 811557 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015442186.1 XP_015297672.1 7 - 8 849 58

View Full Product Details