Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAAR9
Entrez Gene ID
102136054
Gene Name
trace amine associated receptor 9
Gene Aliases
-
Chromosome Location
Chr.4: 36696382 - 36697493 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005595969.1 XP_005596026.1 1 - 1 715 116

View Full Product Details