Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCANP1
Entrez Gene ID
102128054
Gene Name
dendritic cell associated nuclear protein
Gene Aliases
C6H5orf20
Chromosome Location
Chr.6: 134516284 - 134520921 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015451908.1 XP_015307394.1 1 - 1 2172 64

View Full Product Details