Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIPN
Entrez Gene ID
102137951
Gene Name
lipase family member N
Gene Aliases
-
Chromosome Location
Chr.9: 87315929 - 87334281 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005565891.2 XP_005565948.1 2 - 3 290 93

View Full Product Details