Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNA7
Entrez Gene ID
102140355
Gene Name
potassium voltage-gated channel subfamily A member 7
Gene Aliases
-
Chromosome Location
Chr.19: 49882922 - 49889101 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589853.2 XP_005589910.2 1 - 2 942 102

View Full Product Details