Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBARP
Entrez Gene ID
102145612
Gene Name
CACN subunit beta associated regulatory protein
Gene Aliases
C19H19orf26
Chromosome Location
Chr.19: 1005015 - 1014055 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015440173.1 XP_015295659.1 4 - 5 563 92

View Full Product Details