Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A44
Entrez Gene ID
102120541
Gene Name
solute carrier family 25 member 44
Gene Aliases
-
Chromosome Location
Chr.1: 95519182 - 95540493 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541471.2 XP_005541528.1 2 - 3 947 66

View Full Product Details