Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HTR6
Entrez Gene ID
102137033
Gene Name
5-hydroxytryptamine receptor 6
Gene Aliases
-
Chromosome Location
Chr.1: 208747067 - 208762442 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005544575.2 XP_005544632.1 2 - 3 1497 81
GenBank mRNA AY650371.1 - 2 - 3 216 81

View Full Product Details